Anti-p40 Rabbit Polyclonal Antibody
Catalog # 76960-082
Supplier: ANTIBODIES.COM LLC
Specifications
- Antibody Type:Primary
- Antigen Name:40 kDa Rab9 effector protein
- Antigen Symbol:p40
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# Q9H3D4
- Antigen Synonyms:OTTMUSP00000012899|DKFZp686P1077|OTTMUSP00000012898|OTTHUMP00000022126|8430412M01Rik|9530020d24rik|AV073337|bA65N13.1|C87311
- Storage Buffer:Supplied in 10 mM Phosphate Buffered Saline; without Sodium Azide and carrier free
- Molecular Weight:40 kDa
- Storage Temperature:Shipped at 4 ℃. Upon delivery aliquot and store at –20 ℃. Avoid freeze/thaw cycles.
- Concentration:1 mg/ml
- Shipping Temperature:Shipped on blue ice at 4 ℃
- Immunogen:Recombinant fragment of human p40 protein
- Tested Applications:IHC-P
- Purification:Protein A chromatography
- Cat. No.:76960-082
- Packaging:Plastic vial
Specifications
About this item
Rabbit polyclonal antibody to p40 for IHC-P with samples derived from Human.
- Validated Applications: IHC-P
- Recommended Dilutions: IHC-P: 1 to 2 µg/ml
Type: Primary
Antigen: p40
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human