Anti-Mast Cell Tryptase Mouse Monoclonal Antibody [clone: TPSAB1/1963]
76957-690
- Antibody Type:Primary
- Antigen Name:alpha II
- Antigen Symbol:Mast Cell Tryptase
- Clonality:Monoclonal
- Clone:TPSAB1/1963
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1 kappa
- Reactivity:Human
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# Q15661
- Antigen Synonyms:MCP 7|Lung tryptase|Skin tryptase|Mast cell alpha II tryptase|Mast cell beta I tryptase|Pituitary tryptase|Mast cell protease 7|Mast cell protease II|Mast Cell Tryptase
- Storage Buffer:Supplied in 10 mM Phosphate Buffered Saline; without Sodium Azide and carrier free
- Molecular Weight:31 - 36 kDa
- Storage Temperature:Shipped at 4 ℃. Upon delivery aliquot and store at –20 ℃. Avoid freeze/thaw cycles.
- Concentration:1 mg/ml
- Shipping Temperature:Shipped on blue ice at 4 ℃
- Immunogen:Recombinant fragment, around amino acids 115 - 223, of human Tryptase protein. The exact sequence is proprietary
- Tested Applications:IHC-P
- Purification:Protein A/G chromatography
- Cat. No.:76957-690
- Packaging:Plastic vial
Mouse monoclonal [TPSAB1/1963] antibody to Mast Cell Tryptase for IHC-P with samples derived from Human.
- Validated Applications: IHC-P
- Recommended Dilutions: IHC-P: 1 to 2 µg/ml
Type: Primary
Antigen: Mast Cell Tryptase
Clonality: Monoclonal
Clone: TPSAB1/1963
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human