Order Entry
United States
ContactUsLinkComponent
Anti-Melanoma gp100 Rabbit Recombinant Antibody [clone: PMEL/1825R]
Anti-Melanoma gp100 Rabbit Recombinant Antibody [clone: PMEL/1825R]
Catalog # 76956-712
Supplier:  Antibodies.com
Anti-Melanoma gp100 Rabbit Recombinant Antibody [clone: PMEL/1825R]
Catalog # 76956-712
Supplier:  Antibodies.com
Supplier Number:  A253156-100

Specifications

  • Size:
    100 µg
  • Cat. No.:
    76956-712

Specifications

About this item

Recombinant rabbit monoclonal [PMEL/1825R] antibody to Melanoma gp100 for WB and IHC-P with samples derived from Human.

  • Validated Applications: WB, IHC-P
  • Recommended Dilutions: WB: 2 to 4 µg/ml, IHC-P: 1 to 2 µg/ml

Type: Primary
Antigen: Melanoma gp100
Clonality: Recombinant
Clone: PMEL/1825R
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human