Anti-p75 NGF Receptor Mouse Monoclonal Antibody [clone: NGFR5]
Catalog # 76950-552
Supplier: ANTIBODIES.COM LLC
Specifications
- Antibody Type:Primary
- Antigen Name:CD271
- Antigen Symbol:p75 NGF Receptor
- Clonality:Monoclonal
- Clone:NGFR5
- Conjugation:Unconjugated
- Flow Cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG1 kappa
- Reactivity:Baboon,Rabbit,Monkey,Ferret,Human,Cat
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# P08138
- Antigen Synonyms:CD271 antigen|Low affinity neurotrophin receptor p75NTR|Nerve growth factor receptor TNFR superfamily member 16|NGF receptor|p75 ICD|Gp80-LNGFR|Low affinity nerve growth factor receptor|Low-affinity nerve growth factor receptor|Gp80 LNGFR
- Storage Buffer:Supplied in 10 mM Phosphate Buffered Saline; without Sodium Azide and carrier free
- Molecular Weight:75 kDa
- Storage Temperature:Shipped at 4 ℃. Upon delivery aliquot and store at –20 ℃. Avoid freeze/thaw cycles.
- Concentration:1 mg/ml
- Shipping Temperature:Shipped on blue ice at 4 ℃
- Immunogen:NGFR from A875 melanoma cells
- Tested Applications:IHC-P
- Purification:Protein A/G chromatography
- Cat. No.:76950-552
- Packaging:Plastic vial
Specifications
About this item
Mouse monoclonal [NGFR5] antibody to p75 NGF Receptor for Flow Cytometry, IF and IHC-P with samples derived from Human, Monkey, Baboon, Feline, Rabbit and Ferret.
- Validated Applications: Flow Cytometry, IF, IHC-P
- Recommended Dilutions: Flow Cytometry: 1 to 2 µg/million cells, IF: 1 to 2 µg/ml, IHC-P: 1 to 2 µg/ml
Type: Primary
Antigen: p75 NGF Receptor
Clonality: Monoclonal
Clone: NGFR5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human;Monkey;Baboon;Cat;Rabbit;Ferret