Anti-Cytokeratin 6A Mouse Recombinant Antibody [clone: rKRT6A/2100]
76955-940
- Antibody Type:Primary
- Antigen Name:CK-6A
- Antigen Symbol:Cytokeratin 6A
- Clonality:Recombinant
- Clone:rKRT6A/2100
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1 kappa
- Reactivity:Human
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# P02538
- Antigen Synonyms:K2C6A_HUMAN|Cytokeratin 6a|Keratin|CK6A|CK6C|Cytokeratin-6A|Cytokeratin-6D|CK-6D|CK6D
- Storage Buffer:Supplied in 10 mM Phosphate Buffered Saline; without Sodium Azide and carrier free
- Molecular Weight:58 kDa
- Storage Temperature:Shipped at 4 ℃. Upon delivery aliquot and store at –20 ℃. Avoid freeze/thaw cycles.
- Concentration:1 mg/ml
- Shipping Temperature:Shipped on blue ice at 4 ℃
- Immunogen:Recombinant full-length human Cytokeratin 6A protein
- Tested Applications:IHC-P
- Purification:Protein A/G chromatography
- Cat. No.:76955-940
- Packaging:Plastic vial
Recombinant mouse monoclonal [rKRT6A/2100] antibody to Cytokeratin 6A for IHC-P with samples derived from Human.
- Validated Applications: IHC-P
- Recommended Dilutions: IHC-P: 1 to 2 µg/ml
Type: Primary
Antigen: Cytokeratin 6A
Clonality: Recombinant
Clone: rKRT6A/2100
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human