Anti-beta Amyloid Mouse Monoclonal Antibody [clone: APP/3345]
Catalog # 76959-760
Supplier: ANTIBODIES.COM LLC
Specifications
- Antibody Type:Primary
- Antigen Name:A4_HUMAN
- Antigen Symbol:beta Amyloid
- Clonality:Monoclonal
- Clone:APP/3345
- Conjugation:Unconjugated
- Flow Cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG2c kappa
- Reactivity:Human
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# P05067
- Antigen Synonyms:AICD-59|ABETA|AD1|ABPP|AID(50)|AICD-50|AICD-57|AID(57)|AAA
- Storage Buffer:Supplied in 10 mM Phosphate Buffered Saline; without Sodium Azide and carrier free
- Molecular Weight:4 - 46 kDa (beta - Amyloid)/100 - 125 kDa (Amyloid A4)
- Storage Temperature:Shipped at 4 ℃. Upon delivery aliquot and store at –20 ℃. Avoid freeze/thaw cycles.
- Concentration:1 mg/ml
- Shipping Temperature:Shipped on blue ice at 4 ℃
- Immunogen:Recombinant fragment, around amino acids 578 - 680, of human beta Amyloid protein. The exact sequence is proprietary
- Tested Applications:IHC-P
- Purification:Protein A chromatography
- Cat. No.:76959-760
- Packaging:Plastic vial
Specifications
About this item
Mouse monoclonal [APP/3345] antibody to beta Amyloid for IF, Flow Cytometry and IHC-P with samples derived from Human.
- Validated Applications: IF, Flow Cytometry, IHC-P
- Recommended Dilutions: IF: 1 to 2 µg/ml, Flow Cytometry: 1 to 2 µg/million cells, IHC-P: 1 to 2 µg/ml
Type: Primary
Antigen: beta Amyloid
Clonality: Monoclonal
Clone: APP/3345
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2c kappa
Reactivity: Human