Anti-HSP60 Rabbit Recombinant Antibody [clone: HSPD1/2206R]
Catalog # 76965-536
Supplier: ANTIBODIES.COM LLC
Specifications
- Antibody Type:Primary
- Antigen Name:60 kDa chaperonin
- Antigen Symbol:HSP60
- Clonality:Recombinant
- Clone:HSPD1/2206R
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG kappa
- Reactivity:Hamster,Pig,Rabbit,Monkey,Sheep,Chicken,Human,Bovine,Dog,Rat,Mouse
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# P10809
- Antigen Synonyms:Chaperonin, 60-KD|GROEL|Heat shock 60kDa protein 1|Chaperonin 60|CH60_HUMAN|fa04a05|CPN60|60 kDa heat shock protein, mitochondrial|heat shock 60kDa protein 1 (chaperonin)
- Storage Buffer:Supplied in 10 mM Phosphate Buffered Saline; without Sodium Azide and carrier free
- Molecular Weight:60 kDa
- Storage Temperature:Shipped at 4 ℃. Upon delivery aliquot and store at –20 ℃. Avoid freeze/thaw cycles.
- Concentration:1 mg/ml
- Shipping Temperature:Shipped on blue ice at 4 ℃
- Immunogen:Recombinant full-length human HSP60 protein
- Tested Applications:IHC-P
- Purification:Protein A/G chromatography
- Cat. No.:76965-536
- Packaging:Plastic vial
Specifications
About this item
Recombinant rabbit monoclonal [HSPD1/2206R] antibody to HSP60 for WB and IHC-P with samples derived from Human, Mouse, Rat, Hamster, Sheep, Rabbit, Bovine, Canine, Porcine, Monkey and Chicken.
- Validated Applications: WB, IHC-P
- Recommended Dilutions: WB: 1 to 2 µg/ml, IHC-P: 1 to 2 µg/ml
Type: Primary
Antigen: HSP60
Clonality: Recombinant
Clone: HSPD1/2206R
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG kappa
Reactivity: Human;Mouse;Rat;Hamster;Sheep;Rabbit;Bovine;Dog;Porcine;Monkey;Chicken