Anti-ALK Rabbit Recombinant Antibody [clone: ALK1/2766R]
Catalog # 76954-952
Supplier: ANTIBODIES.COM LLC
Specifications
- Antibody Type:Primary
- Antigen Name:ALK
- Antigen Symbol:ALK
- Clonality:Recombinant
- Clone:ALK1/2766R
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# Q9UM73
- Antigen Synonyms:ALK/EML4 fusion gene, included|anaplastic lymphoma kinase (Ki-1)|CD 246|ALK/NPM1 fusion gene, included|Anaplastic lymphoma kinase|anaplastic lymphoma receptor tyrosine kinase|ALK tyrosine kinase receptor|ALK_HUMAN|Anaplastic lymphoma kinase Ki1
- Storage Buffer:Supplied in 10 mM Phosphate Buffered Saline; without Sodium Azide and carrier free
- Molecular Weight:80 (hybrid)/200 kDa (wild type)
- Storage Temperature:Shipped at 4 ℃. Upon delivery aliquot and store at –20 ℃. Avoid freeze/thaw cycles.
- Concentration:1 mg/ml
- Shipping Temperature:Shipped on blue ice at 4 ℃
- Immunogen:Recombinant full-length human ALK protein
- Tested Applications:IHC-P
- Purification:Protein A/G chromatography
- Cat. No.:76954-952
- Packaging:Plastic vial
Specifications
About this item
Recombinant rabbit monoclonal [ALK1/2766R] antibody to ALK for IHC-P with samples derived from Human.
- Validated Applications: IHC-P
- Recommended Dilutions: IHC-P: 1 to 2 µg/ml
Type: Primary
Antigen: ALK
Clonality: Recombinant
Clone: ALK1/2766R
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human