Anti-Estrogen Receptor alpha Mouse Monoclonal Antibody [clone: ESR1/3559]
Catalog # 76960-854
Supplier: ANTIBODIES.COM LLC
Specifications
- Antibody Type:Primary
- Antigen Name:DKFZp686N23123
- Antigen Symbol:Estrogen Receptor alpha
- Clonality:Monoclonal
- Clone:ESR1/3559
- Conjugation:Unconjugated
- Flow Cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG2b kappa
- Reactivity:Human
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# P03372
- Antigen Synonyms:Estradiol receptor|ER|Era|ER alpha|ESRA|ER-alpha|ESR|ESR1|ESR1_HUMAN
- Storage Buffer:Supplied in 10 mM Phosphate Buffered Saline; without Sodium Azide and carrier free
- Molecular Weight:~67 kDa
- Storage Temperature:Shipped at 4 ℃. Upon delivery aliquot and store at –20 ℃. Avoid freeze/thaw cycles.
- Concentration:1 mg/ml
- Shipping Temperature:Shipped on blue ice at 4 ℃
- Immunogen:Recombinant fragment, around amino acids 129 - 312, of human ESR1 protein. The exact sequence is proprietary
- Tested Applications:IHC-P
- Purification:Protein A/G chromatography
- Cat. No.:76960-854
- Packaging:Plastic vial
Specifications
About this item
Mouse monoclonal [ESR1/3559] antibody to Estrogen Receptor alpha for Flow Cytometry and IHC-P with samples derived from Human.
- Validated Applications: Flow Cytometry, IHC-P
- Recommended Dilutions: Flow Cytometry: 1 to 2 µg/million cells, IHC-P: 1 to 2 µg/ml
Type: Primary
Antigen: Estrogen Receptor alpha
Clonality: Monoclonal
Clone: ESR1/3559
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity: Human