Anti-HER4 Mouse Monoclonal Antibody [clone: ERBB4/2581]
Catalog # 76962-164
Supplier: ANTIBODIES.COM LLC
Specifications
- Antibody Type:Primary
- Antigen Name:ERBB4_HUMAN
- Antigen Symbol:HER4
- Clonality:Monoclonal
- Clone:ERBB4/2581
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1 kappa
- Reactivity:Human
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# Q15303
- Antigen Synonyms:Avian erythroblastic leukemia viral oncogene homolog 4|EC 2.7.10.1|Avian erythroblastic leukemia viral v erb b2 oncogene homolog 4|Erbb4|ALS19|ERBB4 intracellular domain|E4ICD|4ICD|ErbB 4
- Storage Buffer:Supplied in 10 mM Phosphate Buffered Saline; without Sodium Azide and carrier free
- Molecular Weight:180 kDa (precursor)/80/120 kDa (cleaved)
- Storage Temperature:Shipped at 4 ℃. Upon delivery aliquot and store at –20 ℃. Avoid freeze/thaw cycles.
- Concentration:1 mg/ml
- Shipping Temperature:Shipped on blue ice at 4 ℃
- Immunogen:Recombinant fragment, around amino acids 1116 - 1269, of human ERBB4 protein. The exact sequence is proprietary
- Tested Applications:IHC-P
- Purification:Protein A/G chromatography
- Cat. No.:76962-164
- Packaging:Plastic vial
Specifications
About this item
Mouse monoclonal [ERBB4/2581] antibody to HER4 for IHC-P with samples derived from Human.
- Validated Applications: IHC-P
- Recommended Dilutions: IHC-P: 1 to 2 µg/ml
Type: Primary
Antigen: HER4
Clonality: Monoclonal
Clone: ERBB4/2581
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human