Anti-Cathepsin D Mouse Monoclonal Antibody [clone: CTSD/2781]
Catalog # 76962-612
Supplier: ANTIBODIES.COM LLC
Specifications
- Antibody Type:Primary
- Antigen Name:CatD
- Antigen Symbol:Cathepsin D
- Clonality:Monoclonal
- Clone:CTSD/2781
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1 kappa
- Reactivity:Human
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# P07339
- Antigen Synonyms:Epididymis secretory sperm binding protein Li 130P|Cathepsin D heavy chain|CTSD|CATD_HUMAN|CD|CPSD|CLN10|Cathepsin D|Ceroid lipofuscinosis neuronal 10
- Storage Buffer:Supplied in 10 mM Phosphate Buffered Saline; without Sodium Azide and carrier free
- Molecular Weight:48 kDa (Procathepsin D)/34 kDa (Cathepsin D)/14 kDa (Cathepsin D)
- Storage Temperature:Shipped at 4 ℃. Upon delivery aliquot and store at –20 ℃. Avoid freeze/thaw cycles.
- Concentration:1 mg/ml
- Shipping Temperature:Shipped on blue ice at 4 ℃
- Immunogen:Recombinant fragment, around amino acids 104 - 250, of human Cathepsin D protein. The exact sequence is proprietary
- Tested Applications:IHC-P
- Purification:Protein A/G chromatography
- Cat. No.:76962-612
- Packaging:Plastic vial
Specifications
About this item
Mouse monoclonal [CTSD/2781] antibody to Cathepsin D for WB and IHC-P with samples derived from Human.
- Validated Applications: WB, IHC-P
- Recommended Dilutions: WB: 1 to 2 µg/ml, IHC-P: 1 to 2 µg/ml
Type: Primary
Antigen: Cathepsin D
Clonality: Monoclonal
Clone: CTSD/2781
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human