Anti-Uroplakin 1A Mouse Monoclonal Antibody [clone: UPK1A/2921]
Catalog # 76950-490
Supplier: ANTIBODIES.COM LLC
Specifications
- Antibody Type:Primary
- Antigen Symbol:Uroplakin 1A
- Clonality:Monoclonal
- Clone:UPK1A/2921
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG2b kappa
- Reactivity:Human
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# O00322
- Storage Buffer:Supplied in 10 mM Phosphate Buffered Saline; without Sodium Azide and carrier free
- Molecular Weight:29 kDa
- Storage Temperature:Shipped at 4 ℃. Upon delivery aliquot and store at –20 ℃. Avoid freeze/thaw cycles.
- Concentration:1 mg/ml
- Shipping Temperature:Shipped on blue ice at 4 ℃
- Immunogen:Recombinant fragment, around amino acids 114 - 173, of human Uroplakin 1A protein. The exact sequence is proprietary
- Tested Applications:IHC-P
- Purification:Protein A/G chromatography
- Cat. No.:76950-490
- Packaging:Plastic vial
Specifications
About this item
Mouse monoclonal [UPK1A/2921] antibody to Uroplakin 1A for ELISA and IHC-P with samples derived from Human.
- Validated Applications: ELISA, IHC-P
- Recommended Dilutions: IHC-P: 1 to 2 µg/ml
Type: Primary
Antigen: Uroplakin 1A
Clonality: Monoclonal
Clone: UPK1A/2921
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity: Human