Order Entry
United States
ContactUsLinkComponent
Anti-Mesothelin Mouse Monoclonal Antibody [clone: MSLN/2131] (Biotin)
Anti-Mesothelin Mouse Monoclonal Antibody [clone: MSLN/2131] (Biotin)
Catalog # 76963-880
Supplier:  ANTIBODIES.COM LLC
undefined
Anti-Mesothelin Mouse Monoclonal Antibody [clone: MSLN/2131] (Biotin)
Catalog # 76963-880
Supplier:  ANTIBODIES.COM LLC
Supplier Number:  A251214-50

Specifications

  • Size:
    50 µg
  • Cat. No.:
    76963-880

Specifications

About this item

Mouse monoclonal [MSLN/2131] antibody to Mesothelin (Biotin) for IHC-P with samples derived from Human, Mouse and Rat.

  • Validated Applications: IHC-P
  • Recommended Dilutions: IHC-P: 2 to 4 µg/ml

Type: Primary
Antigen: Mesothelin
Clonality: Monoclonal
Clone: MSLN/2131
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity: Human;Mouse;Rat