Anti-CD19 Mouse Recombinant Antibody [clone: rCD19/4591]
Catalog # 76960-444
Supplier: ANTIBODIES.COM LLC
Specifications
- Antibody Type:Primary
- Antigen Name:Antibody deficiency due to defect in CD19, included
- Antigen Symbol:CD19
- Clonality:Recombinant
- Clone:rCD19/4591
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG2a kappa
- Reactivity:Human
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# P15391
- Antigen Synonyms:CD19 antigen|Cd19 protein|AW495831|B lymphocyte surface antigen B4|B-lymphocyte antigen CD19|B lymphocyte antigen CD19|CD19 molecule|CD19_HUMAN|B-lymphocyte surface antigen B4
- Storage Buffer:Supplied in 10 mM Phosphate Buffered Saline with 0.05% BSA and 0.05% Sodium Azide
- Molecular Weight:95 kDa
- Storage Temperature:Shipped at 4 ℃. Upon delivery aliquot and store at –20 ℃. Avoid freeze/thaw cycles.
- Concentration:200 µg/ml
- Shipping Temperature:Shipped on blue ice at 4 ℃
- Immunogen:Synthetic peptide corresponding to amino acids within 456 - 556 of CD19 protein
- Tested Applications:IHC-P
- Purification:Protein A/G chromatography
- Cat. No.:76960-444
- Packaging:Plastic vial
Specifications
About this item
Recombinant mouse monoclonal [rCD19/4591] antibody to CD19 for IHC-P with samples derived from Human.
- Validated Applications: IHC-P
- Recommended Dilutions: IHC-P: 1 to 2 µg/ml
Type: Primary
Antigen: CD19
Clonality: Recombinant
Clone: rCD19/4591
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Human