Order Entry
United States
ContactUsLinkComponent
Anti-MSK1 Mouse Monoclonal Antibody [clone: PCRP-RPS6KA5-1A8]
Anti-MSK1 Mouse Monoclonal Antibody [clone: PCRP-RPS6KA5-1A8]
Catalog # 76962-712
Supplier:  ANTIBODIES.COM LLC
undefined
Anti-MSK1 Mouse Monoclonal Antibody [clone: PCRP-RPS6KA5-1A8]
Catalog # 76962-712
Supplier:  ANTIBODIES.COM LLC
Supplier Number:  A250552-100

Specifications

  • Size:
    100 µg
  • Cat. No.:
    76962-712

Specifications

About this item

Mouse monoclonal [PCRP-RPS6KA5-1A8] antibody to MSK1 for Flow Cytometry, IF and WB with samples derived from Human.

  • Validated Applications: Flow Cytometry, IF, WB
  • Recommended Dilutions: Flow Cytometry: 1 to 2 µg/million cells, IF: 1 to 2 µg/ml, WB: 1 to 2 µg/ml

Type: Primary
Antigen: MSK1
Clonality: Monoclonal
Clone: PCRP-RPS6KA5-1A8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human