Anti-Sarcomeric Actinin alpha 2 Mouse Monoclonal Antibody [clone: ACTN2/3292]
76956-812
- Size:100 µg
- Cat. No.:76956-812
Mouse monoclonal [ACTN2/3292] antibody to Sarcomeric Actinin alpha 2 for WB and IHC-P with samples derived from Human.
- Validated Applications: WB, IHC-P
- Recommended Dilutions: WB: 1 to 2 µg/ml, IHC-P: 1 to 2 µg/ml
Type: Primary
Antigen: Sarcomeric Actinin alpha 2
Clonality: Monoclonal
Clone: ACTN2/3292
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity: Human