Anti-CD84 Mouse Monoclonal Antibody [clone: 153-4D9]
Catalog # 76952-994
Supplier: ANTIBODIES.COM LLC
Specifications
- Antibody Type:Primary
- Antigen Name:CD84
- Antigen Symbol:CD84
- Clonality:Monoclonal
- Clone:153-4D9
- Conjugation:Unconjugated
- Flow Cytometry:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG1 kappa
- Reactivity:Human
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# Q9UI8
- Antigen Synonyms:Hly9 beta|hCD84|Cell surface antigen MAX.3|Hly9-beta|CDw84|Leucocyte differentiation antigen CD84|CD84 molecule|CD84 antigen|CD_antigen=CD84
- Storage Buffer:Supplied in 10 mM Phosphate Buffered Saline with 0.05% BSA and 0.05% Sodium Azide
- Molecular Weight:74 kDa
- Storage Temperature:Shipped at 4 ℃. Upon delivery aliquot and store at –20 ℃. Avoid freeze/thaw cycles.
- Concentration:200 µg/ml
- Shipping Temperature:Shipped on blue ice at 4 ℃
- Immunogen:MLR-stimulated lymphocytes
- Purification:Protein A/G chromatography
- Cat. No.:76952-994
- Packaging:Plastic vial
Specifications
About this item
Mouse monoclonal [153-4D9] antibody to CD84 for Flow Cytometry and IF with samples derived from Human.
- Validated Applications: Flow Cytometry, IF
- Recommended Dilutions: Flow Cytometry: 1 to 2 µg/million cells, IF: 1 to 2 µg/ml
Type: Primary
Antigen: CD84
Clonality: Monoclonal
Clone: 153-4D9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human