Order Entry
United States
ContactUsLinkComponent
 
Anti-INSM2 Mouse Monoclonal Antibody [clone: INSM2/4291]
Catalog # 76958-904
Supplier:  ANTIBODIES.COM LLC
undefined
Anti-INSM2 Mouse Monoclonal Antibody [clone: INSM2/4291]
Catalog # 76958-904
Supplier:  ANTIBODIES.COM LLC
Supplier Number:  A250418-100

Specifications

  • Size:
    100 µg
  • Cat. No.:
    76958-904

Specifications

About this item

Mouse monoclonal [INSM2/4291] antibody to INSM2 for IHC-P with samples derived from Human.

  • Validated Applications: IHC-P
  • Recommended Dilutions: IHC-P: 1 to 2 µg/ml

Type: Primary
Antigen: INSM2
Clonality: Monoclonal
Clone: INSM2/4291
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human