Anti-HNF1 alpha Mouse Monoclonal Antibody [clone: HNF1A/2087]
76962-218
- Antibody Type:Primary
- Antigen Name:Albumin proximal factor
- Antigen Symbol:HNF1 alpha
- Clonality:Monoclonal
- Clone:HNF1A/2087
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1 kappa
- Reactivity:Human
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# P20823
- Antigen Synonyms:Hepatic transcription factor 1|HNF-1A|HNF 1|Hepatic nuclear factor 1|Hepatic transcription factor 1 alpha|HNF 1A|HNF-1-alpha|Hepatocyte nuclear factor 1-alpha|Hepatic nuclear factor 1 alpha
- Storage Buffer:Supplied in 10 mM Phosphate Buffered Saline with 0.05% BSA and 0.05% Sodium Azide
- Molecular Weight:79 kDa
- Storage Temperature:Shipped at 4 ℃. Upon delivery aliquot and store at –20 ℃. Avoid freeze/thaw cycles.
- Concentration:200 µg/ml
- Shipping Temperature:Shipped on blue ice at 4 ℃
- Immunogen:Recombinant fragment, around amino acids 214 - 339, of human HNF1A protein. The exact sequence is proprietary
- Tested Applications:IHC-P
- Purification:Protein A/G chromatography
- Cat. No.:76962-218
- Packaging:Plastic vial
Mouse monoclonal [HNF1A/2087] antibody to HNF1 alpha for ELISA and IHC-P with samples derived from Human.
- Validated Applications: ELISA, IHC-P
- Recommended Dilutions: IHC-P: 1 to 2 µg/ml
Type: Primary
Antigen: HNF1 alpha
Clonality: Monoclonal
Clone: HNF1A/2087
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human