Anti-beta III Spectrin Mouse Recombinant Antibody [clone: rSPTBN2/1778]
Catalog # 76967-626
Supplier: ANTIBODIES.COM LLC
Specifications
- Antibody Type:Primary
- Antigen Name:Beta III spectrin
- Antigen Symbol:beta III Spectrin
- Clonality:Recombinant
- Clone:rSPTBN2/1778
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Human
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# O15020
- Antigen Synonyms:Spectrin non erythroid beta chain 2|SCAR14|GTRAP41|Spectrin beta non erythrocytic 2|SCA5|Spectrin beta III sigma 2|SCA 5|Glutamate transporter EAAT4 associated protein 41|Spectrin beta chain brain 2
- Storage Buffer:Supplied in 10 mM Phosphate Buffered Saline with 0.05% BSA and 0.05% Sodium Azide
- Molecular Weight:246 kDa
- Storage Temperature:Shipped at 4 ℃. Upon delivery aliquot and store at –20 ℃. Avoid freeze/thaw cycles.
- Concentration:200 µg/ml
- Shipping Temperature:Shipped on blue ice at 4 ℃
- Immunogen:Recombinant fragment, around amino acids 356 - 475, of human SPTBN2 protein. The exact sequence is proprietary
- Purification:Protein A/G chromatography
- Cat. No.:76967-626
- Packaging:Plastic vial
Specifications
About this item
Recombinant mouse monoclonal [rSPTBN2/1778] antibody to beta III Spectrin for ELISA and WB with samples derived from Human.
- Validated Applications: ELISA, WB
- Recommended Dilutions: WB: 1 to 2 µg/ml
Type: Primary
Antigen: beta III Spectrin
Clonality: Recombinant
Clone: rSPTBN2/1778
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human