Order Entry
United States
ContactUsLinkComponent
Anti-Spastin Mouse Monoclonal Antibody [clone: Sp 3G11-1]
Anti-Spastin Mouse Monoclonal Antibody [clone: Sp 3G11-1]
Catalog # 76952-196
Supplier:  ANTIBODIES.COM LLC
Anti-Spastin Mouse Monoclonal Antibody [clone: Sp 3G11-1]
Catalog # 76952-196
Supplier:  ANTIBODIES.COM LLC
Supplier Number:  A250021-100

Specifications

  • Size:
    100 µg
  • Cat. No.:
    76952-196

Specifications

About this item

Mouse monoclonal [Sp 3G11-1] antibody to Spastin for ELISA, IF, WB and IHC-P with samples derived from Human, Mouse and Rat.

  • Validated Applications: ELISA, IF, WB, IHC-P
  • Recommended Dilutions: IF: 1 to 2 µg/ml, WB: 1 to 2 µg/ml, IHC-P: 1 to 2 µg/ml

Type: Primary
Antigen: Spastin
Clonality: Monoclonal
Clone: Sp 3G11-1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Human;Mouse;Rat