Order Entry
United States
ContactUsLinkComponent
 
Anti-Intelectin 1 Mouse Monoclonal Antibody [clone: ITLN1/4061]
Catalog # 76965-634
Supplier:  ANTIBODIES.COM LLC
undefined
Anti-Intelectin 1 Mouse Monoclonal Antibody [clone: ITLN1/4061]
Catalog # 76965-634
Supplier:  ANTIBODIES.COM LLC
Supplier Number:  A249715-100

Specifications

  • Size:
    100 µg
  • Cat. No.:
    76965-634

Specifications

About this item

Mouse monoclonal [ITLN1/4061] antibody to Intelectin 1 for IHC-P with samples derived from Human.

  • Validated Applications: IHC-P
  • Recommended Dilutions: IHC-P: 1 to 2 µg/ml

Type: Primary
Antigen: Intelectin 1
Clonality: Monoclonal
Clone: ITLN1/4061
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity: Human