Anti-OCT-2 Mouse Monoclonal Antibody [clone: Oct2/2136]
Catalog # 76958-040
Supplier: ANTIBODIES.COM LLC
Specifications
- Antibody Type:Primary
- Antigen Name:class 2
- Antigen Symbol:OCT-2
- Clonality:Monoclonal
- Clone:Oct2/2136
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1 kappa
- Reactivity:Human
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# P09086
- Antigen Synonyms:Octamer-binding protein 2|45201|NF A2|OTF-2|Lymphoid restricted immunoglobulin octamer binding protein|Lymphoid-restricted immunoglobulin octamer-binding protein NF-A2|Octamer binding transcription factor 2|Octamer-binding transcription factor 2
- Storage Buffer:Supplied in 10 mM Phosphate Buffered Saline with 0.05% BSA and 0.05% Sodium Azide
- Molecular Weight:60 kDa
- Storage Temperature:Shipped at 4 ℃. Upon delivery aliquot and store at –20 ℃. Avoid freeze/thaw cycles.
- Concentration:200 µg/ml
- Shipping Temperature:Shipped on blue ice at 4 ℃
- Immunogen:Recombinant fragment, around amino acids 112 - 297, of human OCT2 protein. The exact sequence is proprietary
- Tested Applications:IHC-P
- Purification:Protein A/G chromatography
- Cat. No.:76958-040
- Packaging:Plastic vial
Specifications
About this item
Mouse monoclonal [Oct2/2136] antibody to OCT-2 for IHC-P with samples derived from Human.
- Validated Applications: IHC-P
- Recommended Dilutions: IHC-P: 1 to 2 µg/ml
Type: Primary
Antigen: OCT-2
Clonality: Monoclonal
Clone: Oct2/2136
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human