Order Entry
United States
ContactUsLinkComponent
Anti-NRF1 Mouse Monoclonal Antibody [clone: NRF1/2609]
Anti-NRF1 Mouse Monoclonal Antibody [clone: NRF1/2609]
Catalog # 76967-456
Supplier:  ANTIBODIES.COM LLC
undefined
Anti-NRF1 Mouse Monoclonal Antibody [clone: NRF1/2609]
Catalog # 76967-456
Supplier:  ANTIBODIES.COM LLC
Supplier Number:  A249533-100

Specifications

  • Size:
    100 µg
  • Cat. No.:
    76967-456

Specifications

About this item

Mouse monoclonal [NRF1/2609] antibody to NRF1 for WB and IHC-P with samples derived from Human.

  • Validated Applications: WB, IHC-P
  • Recommended Dilutions: WB: 1 to 2 µg/ml, IHC-P: 1 to 2 µg/ml

Type: Primary
Antigen: NRF1
Clonality: Monoclonal
Clone: NRF1/2609
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human