Order Entry
United States
ContactUsLinkComponent
Anti-p75 NGF Receptor Rabbit Recombinant Antibody [clone: NGFR/2550R]
Anti-p75 NGF Receptor Rabbit Recombinant Antibody [clone: NGFR/2550R]
Catalog # 76966-604
Supplier:  ANTIBODIES.COM LLC
Anti-p75 NGF Receptor Rabbit Recombinant Antibody [clone: NGFR/2550R]
Catalog # 76966-604
Supplier:  ANTIBODIES.COM LLC
Supplier Number:  A249505-100

Specifications

  • Size:
    100 µg
  • Cat. No.:
    76966-604

Specifications

About this item

Recombinant rabbit monoclonal [NGFR/2550R] antibody to p75 NGF Receptor for WB and IHC-P with samples derived from Human.

  • Validated Applications: WB, IHC-P
  • Recommended Dilutions: WB: 1 to 2 µg/ml, IHC-P: 1 to 2 µg/ml

Type: Primary
Antigen: p75 NGF Receptor
Clonality: Recombinant
Clone: NGFR/2550R
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human