Order Entry
United States
ContactUsLinkComponent
Anti-Myogenin Mouse Monoclonal Antibody [clone: MGN185]
Anti-Myogenin Mouse Monoclonal Antibody [clone: MGN185]
Catalog # 76962-120
Supplier:  ANTIBODIES.COM LLC
undefined
Anti-Myogenin Mouse Monoclonal Antibody [clone: MGN185]
Catalog # 76962-120
Supplier:  ANTIBODIES.COM LLC
Supplier Number:  A249469-100

Specifications

  • Size:
    100 µg
  • Cat. No.:
    76962-120

Specifications

About this item

Mouse monoclonal [MGN185] antibody to Myogenin for IHC-P with samples derived from Human, Mouse, Rat, Feline and Porcine.

  • Validated Applications: IHC-P
  • Recommended Dilutions: IHC-P: 1 to 2 µg/ml

Type: Primary
Antigen: Myogenin
Clonality: Monoclonal
Clone: MGN185
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human;Mouse;Rat;Cat;Porcine