Anti-MYH11 Mouse Monoclonal Antibody [clone: S mMS-1]
Catalog # 76965-238
Supplier: ANTIBODIES.COM LLC
Specifications
- Antibody Type:Primary
- Antigen Symbol:MYH11
- Clonality:Monoclonal
- Clone:SMMS-1
- Conjugation:Unconjugated
- Flow Cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG1 kappa
- Reactivity:Pig,Rabbit,Chicken,Human,Bovine,Guinea pig,Dog,Rat,Cat
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# P35749
- Storage Buffer:Supplied in 10 mM Phosphate Buffered Saline with 0.05% BSA and 0.05% Sodium Azide
- Molecular Weight:205 kDa (MHC - 1)/200 kDa (MHC - 2)
- Storage Temperature:Shipped at 4 ℃. Upon delivery aliquot and store at –20 ℃. Avoid freeze/thaw cycles.
- Concentration:200 µg/ml
- Shipping Temperature:Shipped on blue ice at 4 ℃
- Immunogen:Human uterus extract
- Tested Applications:IHC-P
- Purification:Protein A/G chromatography
- Cat. No.:76965-238
- Packaging:Plastic vial
Specifications
About this item
Mouse monoclonal [SMMS-1] antibody to MYH11 for Flow Cytometry, IF and IHC-P with samples derived from Human, Bovine, Porcine, Canine, Feline, Rabbit, Rat, Guinea Pig and Chicken.
- Validated Applications: Flow Cytometry, IF, IHC-P
- Recommended Dilutions: Flow Cytometry: 1 to 2 µg/million cells, IF: 1 to 2 µg/ml, IHC-P: 1 to 2 µg/ml
Type: Primary
Antigen: MYH11
Clonality: Monoclonal
Clone: S mMS-1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human;Bovine;Porcine;Dog;Cat;Rabbit;Rat;Guinea Pig;Chicken