Anti-Cytokeratin 13 Mouse Monoclonal Antibody [clone: KRT13/2659]
76966-898
- Antibody Type:Primary
- Antigen Name:47 kDa cytokeratin
- Antigen Symbol:Cytokeratin 13
- Clonality:Monoclonal
- Clone:KRT13/2659
- Conjugation:Unconjugated
- Flow Cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG1 kappa
- Reactivity:Human
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# P13646
- Antigen Synonyms:Cytokeratin 13|Ka13|CK-13|K1C13_HUMAN|Krt-1.13|Cytokeratin-13|Krt1-13|keratin type I cytoskeletal 13|CK13
- Storage Buffer:Supplied in 10 mM Phosphate Buffered Saline with 0.05% BSA and 0.05% Sodium Azide
- Molecular Weight:52 kDa
- Storage Temperature:Shipped at 4 ℃. Upon delivery aliquot and store at –20 ℃. Avoid freeze/thaw cycles.
- Concentration:200 µg/ml
- Shipping Temperature:Shipped on blue ice at 4 ℃
- Immunogen:Recombinant full-length human KRT13 protein
- Tested Applications:IHC-P
- Purification:Protein A/G chromatography
- Cat. No.:76966-898
- Packaging:Plastic vial
Mouse monoclonal [KRT13/2659] antibody to Cytokeratin 13 for IF, Flow Cytometry and IHC-P with samples derived from Human.
- Validated Applications: IF, Flow Cytometry, IHC-P
- Recommended Dilutions: IF: 1 to 2 µg/ml, Flow Cytometry: 1 to 2 µg/million cells, IHC-P: 1 to 2 µg/ml
Type: Primary
Antigen: Cytokeratin 13
Clonality: Monoclonal
Clone: KRT13/2659
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human