Order Entry
United States
ContactUsLinkComponent
Anti-Cytokeratin 1 Mouse Monoclonal Antibody [clone: LHK1]
Anti-Cytokeratin 1 Mouse Monoclonal Antibody [clone: LHK1]
Catalog # 76965-580
Supplier:  ANTIBODIES.COM LLC
undefined
Anti-Cytokeratin 1 Mouse Monoclonal Antibody [clone: LHK1]
Catalog # 76965-580
Supplier:  ANTIBODIES.COM LLC
Supplier Number:  A249119-100

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    67 kDa cytokeratin
  • Antigen Symbol:
    Cytokeratin 1
  • Clonality:
    Monoclonal
  • Clone:
    LHK1
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG2a kappa
  • Reactivity:
    Human,
    Rat
  • Size:
    100 µg
  • Form:
    Liquid
  • Gene ID:
    UniprotID# P04264
  • Antigen Synonyms:
    EHK|Epidermolytic hyperkeratosis 1|CK1|EHK1|EPPK|Cytokeratin1|Cytokeratin-1|CK-1|Cytokeratin 1
  • Storage Buffer:
    Supplied in 10 mM Phosphate Buffered Saline with 0.05% BSA and 0.05% Sodium Azide
  • Molecular Weight:
    67 kDa
  • Sequence:
    VRFVSTSYSGVTR
  • Storage Temperature:
    Shipped at 4 ℃. Upon delivery aliquot and store at –20 ℃. Avoid freeze/thaw cycles.
  • Concentration:
    200 µg/ml
  • Shipping Temperature:
    Shipped on blue ice at 4 ℃
  • Immunogen:
    Synthetic peptide corresponding to 13 amino acids from the C terminal of human Cytokeratin 1
  • Tested Applications:
    IHC-P
  • Purification:
    Protein A/G chromatography
  • Cat. No.:
    76965-580
  • Packaging:
    Plastic vial

Specifications

About this item

Mouse monoclonal [LHK1] antibody to Cytokeratin 1 for IHC-P with samples derived from Human and Rat.

  • Validated Applications: IHC-P
  • Recommended Dilutions: IHC-P: 1 to 2 µg/ml

Type: Primary
Antigen: Cytokeratin 1
Clonality: Monoclonal
Clone: LHK1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Human;Rat