Anti-Androgen Receptor Mouse Monoclonal Antibody [clone: AR441]
Catalog # 76967-912
Supplier: ANTIBODIES.COM LLC
Specifications
- Antibody Type:Primary
- Antigen Name:AIS
- Antigen Symbol:Androgen Receptor
- Clonality:Monoclonal
- Clone:AR441
- Conjugation:Unconjugated
- Flow Cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG1 kappa
- Reactivity:Human
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# P10275
- Antigen Synonyms:Androgen Receptor|Androgen receptor (dihydrotestosterone receptor|AR|spinal and bulbar muscular atrophy|Kennedy disease)|testicular feminization|ANDR_HUMAN|androgen receptor splice variant 4b|Androgen nuclear receptor variant 2
- Storage Buffer:Supplied in 10 mM Phosphate Buffered Saline with 0.05% BSA and 0.05% Sodium Azide
- Molecular Weight:110 kDa
- Sequence:STEDTAEYSPFKGGYTK
- Storage Temperature:Shipped at 4 ℃. Upon delivery aliquot and store at –20 ℃. Avoid freeze/thaw cycles.
- Concentration:200 µg/ml
- Shipping Temperature:Shipped on blue ice at 4 ℃
- Immunogen:Synthetic peptide corresponding to amino acids 302 - 318 of human Androgen Receptor
- Tested Applications:IHC-P
- Purification:Protein A/G chromatography
- Cat. No.:76967-912
- Packaging:Plastic vial
Specifications
About this item
Mouse monoclonal [AR441] antibody to Androgen Receptor for Flow Cytometry, IF and IHC-P with samples derived from Human.
- Validated Applications: Flow Cytometry, IF, IHC-P
- Recommended Dilutions: Flow Cytometry: 1 to 2 µg/million cells, IF: 1 to 2 µg/ml, IHC-P: 1 to 2 µg/ml
Type: Primary
Antigen: Androgen Receptor
Clonality: Monoclonal
Clone: AR441
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human