Anti-APOA4 Mouse Monoclonal Antibody [clone: APOA4/3372]
76966-834
- Antibody Type:Primary
- Antigen Name:Apo AIV
- Antigen Symbol:APOA4
- Clonality:Monoclonal
- Clone:APOA4/3372
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG2b kappa
- Reactivity:Human
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# P06727
- Antigen Synonyms:MGC142156|Apolipoprotein A4|APOA 4|APOA4|MGC142154|ApoA IV|Apolipoprotein A IV
- Storage Buffer:Supplied in 10 mM Phosphate Buffered Saline with 0.05% BSA and 0.05% Sodium Azide
- Molecular Weight:46 kDa
- Storage Temperature:Shipped at 4 ℃. Upon delivery aliquot and store at –20 ℃. Avoid freeze/thaw cycles.
- Concentration:200 µg/ml
- Shipping Temperature:Shipped on blue ice at 4 ℃
- Immunogen:Recombinant fragment, around amino acids 239 - 371, of human APOA4 protein. The exact sequence is proprietary
- Tested Applications:IHC-P
- Purification:Protein A/G chromatography
- Cat. No.:76966-834
- Packaging:Plastic vial
Mouse monoclonal [APOA4/3372] antibody to APOA4 for IHC-P with samples derived from Human.
- Validated Applications: IHC-P
- Recommended Dilutions: IHC-P: 1 to 2 µg/ml
Type: Primary
Antigen: APOA4
Clonality: Monoclonal
Clone: APOA4/3372
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity: Human