Anti-HIF1 alpha Mouse Monoclonal Antibody [clone: HIF1A/84]
Catalog # 76962-340
Supplier: ANTIBODIES.COM LLC
Specifications
- Antibody Type:Primary
- Antigen Name:ARNT interacting protein
- Antigen Symbol:HIF1 alpha
- Clonality:Monoclonal
- Clone:HIF1A/84
- Conjugation:Unconjugated
- Flow Cytometry:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG2b kappa
- Reactivity:Human
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# Q16665
- Antigen Synonyms:HIF 1alpha|ARNT-interacting protein|Basic-helix-loop-helix-PAS protein MOP1|HIF-1-alpha|HIF-1 alpha|Basic helix loop helix PAS protein MOP1|bHLHe78|HIF 1A|Class E basic helix-loop-helix protein 78
- Storage Buffer:Supplied in 10 mM Phosphate Buffered Saline with 0.05% BSA and 0.05% Sodium Azide
- Molecular Weight:92 - 110 kDa
- Storage Temperature:Shipped at 4 ℃. Upon delivery aliquot and store at –20 ℃. Avoid freeze/thaw cycles.
- Concentration:200 µg/ml
- Shipping Temperature:Shipped on blue ice at 4 ℃
- Immunogen:Recombinant human HIF1 alpha protein
- Purification:Protein A/G chromatography
- Cat. No.:76962-340
- Packaging:Plastic vial
Specifications
About this item
Mouse monoclonal [HIF1A/84] antibody to HIF1 alpha for Flow Cytometry and IF with samples derived from Human.
- Validated Applications: Flow Cytometry, IF
- Recommended Dilutions: Flow Cytometry: 1 to 2 µg/million cells, IF: 1 to 2 µg/ml
Type: Primary
Antigen: HIF1 alpha
Clonality: Monoclonal
Clone: HIF1A/84
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity: Human