Anti-Glycophorin A Rabbit Polyclonal Antibody
Catalog # 76953-944
Supplier: ANTIBODIES.COM LLC
Specifications
- Antibody Type:Primary
- Antigen Name:GlycophorinA
- Antigen Symbol:Glycophorin A
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# P02724
- Antigen Synonyms:GLPA_HUMAN|Glycophorin A, included|Glycophorin A|Glycophorin-A|AI853584|Blood group--MN locus|Glycophorin A (MNS blood group)|CD235a|CD_antigen=CD235a
- Storage Buffer:Supplied in 10 mM Phosphate Buffered Saline with 0.05% BSA and 0.05% Sodium Azide
- Molecular Weight:39 kDa
- Storage Temperature:Shipped at 4 ℃. Upon delivery aliquot and store at –20 ℃. Avoid freeze/thaw cycles.
- Concentration:200 µg/ml
- Shipping Temperature:Shipped on blue ice at 4 ℃
- Immunogen:Recombinant human Glycophorin A protein
- Tested Applications:IHC-P
- Purification:Protein A chromatography
- Cat. No.:76953-944
- Packaging:Plastic vial
Specifications
About this item
Rabbit polyclonal antibody to Glycophorin A for WB and IHC-P with samples derived from Human.
- Validated Applications: WB, IHC-P
- Recommended Dilutions: WB: 1 to 2 µg/ml, IHC-P: 1 to 2 µg/ml
Type: Primary
Antigen: Glycophorin A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human