Anti-Glutathione S Transferase Mu 3 Mouse Monoclonal Antibody [clone: CPTC-GSTMu3-1]
Catalog # 76962-806
Supplier: ANTIBODIES.COM LLC
Specifications
- Antibody Type:Primary
- Antigen Name:brain GST
- Antigen Symbol:Glutathione S Transferase Mu 3
- Clonality:Monoclonal
- Clone:CPTC-GSTMu3-1
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG2a kappa
- Reactivity:Human
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# P21266
- Antigen Synonyms:glutathione S transferase M3|glutathione S transferase M3 (brain)|GST class mu 3|glutathione S transferase, Mu 3|Glutathione S-transferase Mu 3|glutathione S aryltransferase M3|glutathione S alkyltransferase M3|brain type mu glutathione S transferase|Glutathione S transferase Mu 3
- Storage Buffer:Supplied in 10 mM Phosphate Buffered Saline with 0.05% BSA and 0.05% Sodium Azide
- Molecular Weight:26 kDa
- Storage Temperature:Shipped at 4 ℃. Upon delivery aliquot and store at –20 ℃. Avoid freeze/thaw cycles.
- Concentration:200 µg/ml
- Shipping Temperature:Shipped on blue ice at 4 ℃
- Immunogen:Recombinant full-length human GSTM3 protein
- Tested Applications:IHC-P
- Purification:Protein A/G chromatography
- Cat. No.:76962-806
- Packaging:Plastic vial
Specifications
About this item
Mouse monoclonal [CPTC-GSTMu3-1] antibody to Glutathione S Transferase Mu 3 for IHC-P with samples derived from Human.
- Validated Applications: IHC-P
- Recommended Dilutions: IHC-P: 1 to 2 µg/ml
Type: Primary
Antigen: Glutathione S Transferase Mu 3
Clonality: Monoclonal
Clone: CPTC-GSTMu3-1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Human