Anti-Glutathione S Transferase Mu 1 Mouse Monoclonal Antibody [clone: CPTC-GSTMu1-3]
76954-772
- Antibody Type:Primary
- Antigen Name:Glutathione S alkyltransferase M1
- Antigen Symbol:Glutathione S Transferase Mu 1
- Clonality:Monoclonal
- Clone:CPTC-GSTMu1-3
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1 kappa
- Reactivity:Human
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# P09488
- Antigen Synonyms:GSTM1 + GSTM2|GTM1|Glutathione S transferase Mu 2|Glutathione S alkyltransferase M2|Glutathione S transferase M1|GTH4|Glutathione S transferase Mu 1|GST class mu 1|GSTM
- Storage Buffer:Supplied in 10 mM Phosphate Buffered Saline with 0.05% BSA and 0.05% Sodium Azide
- Molecular Weight:predicted: 26 kDa/observed: 33 kDa
- Storage Temperature:Shipped at 4 ℃. Upon delivery aliquot and store at –20 ℃. Avoid freeze/thaw cycles.
- Concentration:200 µg/ml
- Shipping Temperature:Shipped on blue ice at 4 ℃
- Immunogen:Recombinant full-length human GSTMu1 protein
- Tested Applications:IHC-P
- Purification:Protein A/G chromatography
- Cat. No.:76954-772
- Packaging:Plastic vial
Mouse monoclonal [CPTC-GSTMu1-3] antibody to Glutathione S Transferase Mu 1 for WB and IHC-P with samples derived from Human.
- Validated Applications: WB, IHC-P
- Recommended Dilutions: WB: 1 to 2 µg/ml, IHC-P: 1 to 2 µg/ml
Type: Primary
Antigen: Glutathione S Transferase Mu 1
Clonality: Monoclonal
Clone: CPTC-GSTMu1-3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human