Anti-Growth Hormone Mouse Monoclonal Antibody [clone: GH/3155]
Catalog # 76950-448
Supplier: ANTIBODIES.COM LLC
Specifications
- Antibody Type:Primary
- Antigen Name:gH
- Antigen Symbol:Growth Hormone
- Clonality:Monoclonal
- Clone:GH/3155
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1 kappa
- Reactivity:Human
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# P01241
- Antigen Synonyms:GH-N|Growth hormone, normal|Growth hormone 1|Growth Hormone|GHB5|Growth hormone B5|GHN|GH1|Growth hormone, pituitary
- Storage Buffer:Supplied in 10 mM Phosphate Buffered Saline with 0.05% BSA and 0.05% Sodium Azide
- Molecular Weight:20 kDa
- Storage Temperature:Shipped at 4 ℃. Upon delivery aliquot and store at –20 ℃. Avoid freeze/thaw cycles.
- Concentration:200 µg/ml
- Shipping Temperature:Shipped on blue ice at 4 ℃
- Immunogen:full-length human Growth Hormone protein
- Tested Applications:IHC-P
- Purification:Protein A/G chromatography
- Cat. No.:76950-448
- Packaging:Plastic vial
Specifications
About this item
Mouse monoclonal [GH/3155] antibody to Growth Hormone for IHC-P with samples derived from Human.
- Validated Applications: IHC-P
- Recommended Dilutions: IHC-P: 1 to 2 µg/ml
Type: Primary
Antigen: Growth Hormone
Clonality: Monoclonal
Clone: GH/3155
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human