Anti-FSH beta Mouse Monoclonal Antibody [clone: SPM107]
Catalog # 76966-926
Supplier: ANTIBODIES.COM LLC
Specifications
- Antibody Type:Primary
- Antigen Symbol:FSH beta
- Clonality:Monoclonal
- Clone:SPM107
- Conjugation:Unconjugated
- Flow Cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG1 kappa
- Reactivity:Human
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# P01225
- Storage Buffer:Supplied in 10 mM Phosphate Buffered Saline with 0.05% BSA and 0.05% Sodium Azide
- Molecular Weight:22 kDa
- Storage Temperature:Shipped at 4 ℃. Upon delivery aliquot and store at –20 ℃. Avoid freeze/thaw cycles.
- Concentration:200 µg/ml
- Shipping Temperature:Shipped on blue ice at 4 ℃
- Immunogen:full-length purified human FSH
- Tested Applications:IHC-P
- Purification:Protein A/G chromatography
- Cat. No.:76966-926
- Packaging:Plastic vial
Specifications
About this item
Mouse monoclonal [SPM107] antibody to FSH beta for Flow Cytometry, IF and IHC-P with samples derived from Human.
- Validated Applications: Flow Cytometry, IF, IHC-P
- Recommended Dilutions: Flow Cytometry: 1 to 2 µg/million cells, IF: 1 to 2 µg/ml, IHC-P: 1 to 2 µg/ml
Type: Primary
Antigen: FSH beta
Clonality: Monoclonal
Clone: SPM107
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human