Anti-Albumin Mouse Monoclonal Antibody [clone: ALB/2142]
Catalog # 76954-428
Supplier: ANTIBODIES.COM LLC
Specifications
- Antibody Type:Primary
- Antigen Name:Growth inhibiting protein 20
- Antigen Symbol:Albumin
- Clonality:Monoclonal
- Clone:ALB/2142
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1 kappa
- Reactivity:Human
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# P02768
- Antigen Synonyms:GIG20|DKFZp779N1935|ALBU_HUMAN|alb|GIG42|Albumin (32 AA)|cell growth inhibiting protein 42|Albumin (AA 34)|Albumin
- Storage Buffer:Supplied in 10 mM Phosphate Buffered Saline with 0.05% BSA and 0.05% Sodium Azide
- Molecular Weight:66 kDa
- Storage Temperature:Shipped at 4 ℃. Upon delivery aliquot and store at –20 ℃. Avoid freeze/thaw cycles.
- Concentration:200 µg/ml
- Shipping Temperature:Shipped on blue ice at 4 ℃
- Immunogen:Recombinant full-length human ALB protein
- Tested Applications:IHC-P
- Purification:Protein A/G chromatography
- Cat. No.:76954-428
- Packaging:Plastic vial
Specifications
About this item
Mouse monoclonal [ALB/2142] antibody to Albumin for ELISA and IHC-P with samples derived from Human.
- Validated Applications: ELISA, IHC-P
- Recommended Dilutions: IHC-P: 1 to 2 µg/ml
Type: Primary
Antigen: Albumin
Clonality: Monoclonal
Clone: ALB/2142
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human