Anti-EPO Mouse Recombinant Antibody [clone: rEPO/1367]
Catalog # 76949-138
Supplier: ANTIBODIES.COM LLC
Specifications
- Antibody Type:Primary
- Antigen Name:EP
- Antigen Symbol:EPO
- Clonality:Recombinant
- Clone:rEPO/1367
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1 kappa
- Reactivity:Human
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# P01588
- Antigen Synonyms:EPO alpha|Epoetin|EPO|Erythropoietin precursor|Erythropoetin|MVCD2
- Storage Buffer:Supplied in 10 mM Phosphate Buffered Saline with 0.05% BSA and 0.05% Sodium Azide
- Molecular Weight:37 kDa
- Storage Temperature:Shipped at 4 ℃. Upon delivery aliquot and store at –20 ℃. Avoid freeze/thaw cycles.
- Concentration:200 µg/ml
- Shipping Temperature:Shipped on blue ice at 4 ℃
- Immunogen:Recombinant fragment, around amino acids 28 - 162, of human EPO protein. The exact sequence is proprietary
- Tested Applications:IHC-P
- Purification:Protein A/G chromatography
- Cat. No.:76949-138
- Packaging:Plastic vial
Specifications
About this item
Recombinant mouse monoclonal [rEPO/1367] antibody to EPO for ELISA and IHC-P with samples derived from Human.
- Validated Applications: ELISA, IHC-P
- Recommended Dilutions: IHC-P: 2 to 4 µg/ml
Type: Primary
Antigen: EPO
Clonality: Recombinant
Clone: rEPO/1367
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human