Anti-Desmin Mouse Monoclonal Antibody [clone: DES/1711]
76959-054
- Antibody Type:Primary
- Antigen Name:CMD1I
- Antigen Symbol:Desmin
- Clonality:Monoclonal
- Clone:DES/1711
- Conjugation:Unconjugated
- Flow Cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG1 kappa
- Reactivity:Hamster,Chicken,Human,Rat,Mouse
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# P17661
- Antigen Synonyms:CSM1|FLJ41013|DESM_HUMAN|DES|Desmin|FLJ12025|FLJ39719|FLJ41793|CSM2
- Storage Buffer:Supplied in 10 mM Phosphate Buffered Saline with 0.05% BSA and 0.05% Sodium Azide
- Molecular Weight:52 kDa
- Storage Temperature:Shipped at 4 ℃. Upon delivery aliquot and store at –20 ℃. Avoid freeze/thaw cycles.
- Concentration:200 µg/ml
- Shipping Temperature:Shipped on blue ice at 4 ℃
- Immunogen:Recombinant human Desmin protein
- Tested Applications:IHC-P
- Purification:Protein A/G chromatography
- Cat. No.:76959-054
- Packaging:Plastic vial
Mouse monoclonal [DES/1711] antibody to Desmin for IF, Flow Cytometry, WB and IHC-P with samples derived from Human, Mouse, Rat, Hamster and Chicken.
- Validated Applications: IF, Flow Cytometry, WB, IHC-P
- Recommended Dilutions: IF: 1 to 2 µg/ml, Flow Cytometry: 1 to 2 µg/million cells, WB: 1 to 2 µg/ml, IHC-P: 1 to 2 µg/ml
Type: Primary
Antigen: Desmin
Clonality: Monoclonal
Clone: DES/1711
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human;Mouse;Rat;Hamster;Chicken