Anti-CFTR Rabbit Recombinant Antibody [clone: CFTR/2290R]
Catalog # 76957-054
Supplier: ANTIBODIES.COM LLC
Specifications
- Antibody Type:Primary
- Antigen Name:ABC 35
- Antigen Symbol:CFTR
- Clonality:Recombinant
- Clone:CFTR/2290R
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# P13569
- Antigen Synonyms:ABC35|ABCC7|ATP-binding cassette sub-family C member 7|cAMP-dependent chloride channel|cAMP dependent chloride channel|ATP binding cassette transporter sub family C member 7|ABCC 7|ATP binding cassette sub family C member 7|ATP Binding Cassette Superfamily C Member 7
- Storage Buffer:Supplied in 10 mM Phosphate Buffered Saline with 0.05% BSA and 0.05% Sodium Azide
- Molecular Weight:165 - 170 kDa
- Storage Temperature:Shipped at 4 ℃. Upon delivery aliquot and store at –20 ℃. Avoid freeze/thaw cycles.
- Concentration:200 µg/ml
- Shipping Temperature:Shipped on blue ice at 4 ℃
- Immunogen:Recombinant full-length human CFTR protein
- Tested Applications:IHC-P
- Purification:Protein A/G chromatography
- Cat. No.:76957-054
- Packaging:Plastic vial
Specifications
About this item
Recombinant rabbit monoclonal [CFTR/2290R] antibody to CFTR for IHC-P with samples derived from Human and Mouse.
- Validated Applications: IHC-P
- Recommended Dilutions: IHC-P: 1 to 2 µg/ml
Type: Primary
Antigen: CFTR
Clonality: Recombinant
Clone: CFTR/2290R
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse