Order Entry
United States
ContactUsLinkComponent
 
Anti-TCR C beta 1 Mouse Monoclonal Antibody [clone: JOVI.1] (APC (Allophycocyanin))
Catalog # 76959-202
Supplier:  Antibodies.com
undefined
Anti-TCR C beta 1 Mouse Monoclonal Antibody [clone: JOVI.1] (APC (Allophycocyanin))
Catalog # 76959-202
Supplier:  Antibodies.com
Supplier Number:  A242886-100

Specifications

  • Size:
    100 tests
  • Cat. No.:
    76959-202

Specifications

About this item

Mouse monoclonal [JOVI.1] antibody to TCR C beta 1 (APC) for Flow Cytometry with samples derived from Human.

  • Validated Applications: Flow Cytometry
  • Recommended Dilutions: Flow Cytometry: This reagent is designed for analysis of human blood cells using 10 µl reagent / 100 µl of whole blood or 106 cells in a suspension.

Type: Primary
Antigen: TCR C beta 1
Clonality: Monoclonal
Clone: JOVI.1
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Human