Anti-CD3 Mouse Monoclonal Antibody [clone: UCHT1] (APC (Allophycocyanin)-Cy7®)
Catalog # 76958-666
Supplier: ANTIBODIES.COM LLC
Specifications
- Antibody Type:Primary
- Antigen Name:CD3 delta
- Antigen Symbol:CD3
- Clonality:Monoclonal
- Clone:UCHT1
- Conjugation:APC (Allophycocyanin)-Cy7®
- Flow Cytometry:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Non-human primates,Human
- Size:100 tests
- Form:Liquid
- Gene ID:UniprotID# P07766
- Antigen Synonyms:CD247 antigen|CD3|CD3 antigen, delta subunit|CD247 antigen, zeta subunit|CD247 molecule|Cd247|4930549J05Rik|A430104F18Rik|AW552088
- Storage Buffer:Supplied in Phosphate Buffered Saline with 15 mM Sodium Azide
- Storage Temperature:Store in the dark at 2...8 ℃. Do not freeze!
- Shipping Temperature:Shipped on blue ice at 4 ℃
- Immunogen:Human thymocytes followed by Sezary T cells
- Purification:This antibody is conjugated with tandem dye APC-Cyanine 7 under optimum conditions The conjugate is purified by size-exclusion chromatography
- Cat. No.:76958-666
- Packaging:Glass bottle
Specifications
About this item
Mouse monoclonal [UCHT1] antibody to CD3 (APC-Cyanine 7) for Flow Cytometry with samples derived from Human and Non-Human Primates.
- Validated Applications: Flow Cytometry
- Recommended Dilutions: Flow Cytometry: This reagent is designed for analysis of human blood cells using 4 µl reagent / 100 µl of whole blood or 106 cells in a suspension.
Type: Primary
Antigen: CD3
Clonality: Monoclonal
Clone: UCHT1
Conjugation: APC (Allophycocyanin)-Cy7®
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human;Non-Human Primates