Anti-Interferon gamma Mouse Monoclonal Antibody [clone: 4S.B3] (PerCP-Cyanine 5.5)
Catalog # 76959-576
Supplier: ANTIBODIES.COM LLC
Specifications
- Antibody Type:Primary
- Antigen Name:IFG
- Antigen Symbol:Interferon gamma
- Clonality:Monoclonal
- Clone:4S.B3
- Conjugation:PerCP (Peridinin-Chlorophyll Protein Complex)-Cy5,5®
- Flow Cytometry:Yes
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Non-human primates,Human
- Size:100 tests
- Form:Liquid
- Gene ID:UniprotID# P01579
- Antigen Synonyms:IFN immune|IFI|IFN, immune|immune interferon|IFN gamma|IFNG|IFN-gamma|Interferon gamma|IFNG_HUMAN
- Storage Buffer:Supplied in Phosphate Buffered Saline with 15 mM Sodium Azide
- Storage Temperature:Store in the dark at 2...8 ℃. Do not freeze!
- Shipping Temperature:Shipped on blue ice at 4 ℃
- Immunogen:Interferon ga mMa derived from human leukocytes
- Purification:This antibody is conjugated with tandem dye PerCP-Cyanine 55 under optimum conditions The conjugate is purified by size-exclusion chromatography
- Cat. No.:76959-576
- Packaging:Glass bottle
Specifications
About this item
Mouse monoclonal [4S.B3] antibody to Interferon gamma (PerCP-Cyanine 5.5) for Flow Cytometry with samples derived from Human and Non-Human Primates.
- Validated Applications: Flow Cytometry
- Recommended Dilutions: Flow Cytometry: This reagent is designed for analysis of human blood cells using 4 µl reagent / 100 µl of whole blood or 106 cells in a suspension.
Type: Primary
Antigen: Interferon gamma
Clonality: Monoclonal
Clone: 4S.B3
Conjugation: PerCP-Cyanine 5.5
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human;Non-Human Primates