To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Symbol:alpha 1 Sodium Potassium ATPase
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Goat
- Isotype:IgG
- Reactivity:Monkey,Human,Dog,Rat,Mouse
- Western Blot:Yes
- Size:300 µg
- Environmentally Preferable:
- Form:Liquid
- Storage Buffer:Supplied in Phosphate Buffered Saline with 20% Glycerol and 0.05% Sodium Azide
- Storage Temperature:Shipped at 4 ℃. Upon delivery aliquot and store at 4 ℃. Avoid freeze/thaw cycles.
- Concentration:3 mg/ml
- Shipping Temperature:Shipped on blue ice at 4 ℃
- Immunogen:Recombinant peptide derived from within residues a.a. 100 to rhe N terminus of human ATP1a1, expressed in and purified from E. coli
- Purification:Affinity purification
- Cat. No.:76834-708
- Packaging:Plastic vial
Specifications
About this item
Anti-alpha 1 Sodium Potassium ATPase antibody for WB with samples derived from Human, Rat, Mouse, Canine and Monkey.
- Validated Applications: WB
- Recommended Dilutions: WB: 1:500 to 1:2000
Type: Primary
Antigen: alpha 1 Sodium Potassium ATPase
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human;Rat;Mouse;Dog;Monkey