Anti-Orc2 Rabbit Polyclonal Antibody
77073-528
- Antibody Type:Primary
- Antigen Name:ORC2
- Antigen Symbol:Orc2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:S. pombe
- Western Blot:Yes
- Size:100 µg
- ChIP:Yes
- Form:Liquid
- Gene ID:UniprotID# Q09142
- Antigen Synonyms:Origin recognition complex subunit 2|Origin recognition complex subunit 2 isoform B|Origin recognition complex, subunit 2, S. cerevisiae, homolog of|ORC2_HUMAN|ORC2L|Origin recognition complex, subunit 2 homolog|Origin of replication 2 (yeast homolog) like|Origin recognition complex subunit 2 like (yeast)|Origin recognition complex protein 2 homolog
- Storage Temperature:Shipped at 4 ℃. Upon delivery aliquot and store at –20 ℃. Avoid freeze/thaw cycles.
- Concentration:2 mg/ml
- Shipping Temperature:Shipped on blue ice at 4 ℃
- Immunogen:Recombinant Schizosaccharomyces pombe Orc2 protein (a.a. 1-175, His tag)
- Purification:Protein A chromatography
- Cat. No.:77073-528
- Packaging:Plastic vial
Rabbit polyclonal antibody to Orc2 for WB, IP and ChIP with samples derived from Schizosaccharomyces pombe.
- Validated Applications: WB, IP, ChIP
- Recommended Dilutions: WB: 1:2000, IP: 1:200
Type: Primary
Antigen: Orc2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Schizosaccharomyces pombe