Order Entry
United States
ContactUsLinkComponent
Anti-Macrophage Infla mMatory Protein 1 beta Rabbit Polyclonal Antibody
Anti-Macrophage Infla mMatory Protein 1 beta Rabbit Polyclonal Antibody
Catalog # 77074-660
Supplier:  ANTIBODIES.COM LLC
undefined
Anti-Macrophage Infla mMatory Protein 1 beta Rabbit Polyclonal Antibody
Catalog # 77074-660
Supplier:  ANTIBODIES.COM LLC
Supplier Number:  A101531-100
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Size:
    100 µg
  • Cat. No.:
    77074-660

Specifications

About this item

Rabbit polyclonal antibody to CCL4 for WB and ELISA with samples derived from Human.

  • Validated Applications: WB, ELISA
  • Recommended Dilutions: WB: 1:500 to 1:1000, ELISA: 1:10000

Type: Primary
Antigen: Macrophage Infla mMatory Protein 1 beta
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human