Anti-MAD1 Rabbit Polyclonal Antibody
Catalog # 77064-340
Supplier: ANTIBODIES.COM LLC
Specifications
- Antibody Type:Primary
- Antigen Name:hMAD1
- Antigen Symbol:MAD1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# Q9Y6D9
- Antigen Synonyms:MAD1L1|MAD1-like protein 1|HsMAD1|Mitotic checkpoint MAD1 protein homolog|MD1L1_HUMAN|Mitotic arrest deficient 1-like protein 1|Mitotic spindle assembly checkpoint protein MAD1|Mitotic arrest deficient 1|MAD1 mitotic arrest deficient like 1 (yeast)
- Storage Buffer:Supplied in Phosphate Buffered Saline (without Mg2+ and Ca2+), pH 7.4, with 150 mM NaCl, 0.02% Sodium Azide, and 50% Glycerol
- Molecular Weight:83 kDa
- Storage Temperature:Shipped at 4 ℃. Upon delivery aliquot and store at –20 ℃. Avoid freeze/thaw cycles.
- Shipping Temperature:Shipped on blue ice at 4 ℃
- Immunogen:Synthetic peptide derived from human MAD1 (amino acids 394 - 443)
- Purification:Purified from rabbit serum by antigen affinity chromatography using the immunizing peptide
- Cat. No.:77064-340
- Packaging:Plastic vial
Specifications
About this item
Rabbit polyclonal antibody to MAD1 for IF and ELISA with samples derived from Human.
- Validated Applications: IF, ELISA
- Recommended Dilutions: IF: 1:100 to 1:500, ELISA: 1:10000
Type: Primary
Antigen: MAD1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human