Anti-A4GNT Rabbit Polyclonal Antibody
Catalog # 77072-930
Supplier: ANTIBODIES.COM LLC
Specifications
- Antibody Type:Primary
- Antigen Name:4-N-acetylglucosaminyltransferase
- Antigen Symbol:A4GNT
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# Q9UNA3
- Antigen Synonyms:Alpha-1|MGC149493|A4GNT|Alpha4GnT|EC 2.4.1.|Alpha 1 4 N acetylglucosaminyltransferase|A4GCT_HUMAN
- Storage Buffer:Supplied in Phosphate Buffered Saline (without Mg2+ and Ca2+), pH 7.4, with 150 mM NaCl, 0.02% Sodium Azide, and 50% Glycerol
- Molecular Weight:39 kDa
- Storage Temperature:Shipped at 4 ℃. Upon delivery aliquot and store at –20 ℃. Avoid freeze/thaw cycles.
- Shipping Temperature:Shipped on blue ice at 4 ℃
- Immunogen:Synthetic peptide derived from human A4GNT (amino acids 31 - 80)
- Purification:Purified from rabbit serum by antigen affinity chromatography using the immunizing peptide
- Cat. No.:77072-930
- Packaging:Plastic vial
Specifications
About this item
Rabbit polyclonal antibody to A4GNT for WB and ELISA with samples derived from Human.
- Validated Applications: WB, ELISA
- Recommended Dilutions: WB: 1:500 to 1:1000, ELISA: 1:40000
Type: Primary
Antigen: A4GNT
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human