Anti-MRP2 Rabbit Polyclonal Antibody
Catalog # 77067-594
Supplier: ANTIBODIES.COM LLC
Specifications
- Antibody Type:Primary
- Antigen Name:ABC30
- Antigen Symbol:MRP2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# Q92887
- Antigen Synonyms:ATP-binding cassette sub-family C member 2|ATP binding cassette subfamily C member 2|Canalicular multidrug resistance protein|CMOAT1|abcC2|cMRP|CMOAT|ATP binding cassette sub family C (CFTR/MRP) member 2|Canalicular multispecific organic anion transporter 1
- Storage Buffer:Supplied in Phosphate Buffered Saline (without Mg2+ and Ca2+), pH 7.4, with 150 mM NaCl, 0.02% Sodium Azide, and 50% Glycerol
- Molecular Weight:174 kDa
- Storage Temperature:Shipped at 4 ℃. Upon delivery aliquot and store at –20 ℃. Avoid freeze/thaw cycles.
- Shipping Temperature:Shipped on blue ice at 4 ℃
- Immunogen:Synthetic peptide derived from human ABCC2 (amino acids 991 - 1040)
- Purification:Purified from rabbit serum by antigen affinity chromatography using the immunizing peptide
- Cat. No.:77067-594
- Packaging:Plastic vial
Specifications
About this item
Rabbit polyclonal antibody to ABCC2 for WB and ELISA with samples derived from Human.
- Validated Applications: WB, ELISA
- Recommended Dilutions: WB: 1:500 to 1:1000, ELISA: 1:10000
Type: Primary
Antigen: MRP2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human